3',5'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4934-1G CA-4934-250M
COBBCA-4934-1GEA 303 EUR
COBBCA-4934-1G COBBCA-4934-250M
3',5'-Dimethoxybiphenyl-3-carbonsäure
3',5'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
MDL: MFCD15143527
CAS-Nummer: 1215205-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR