Terbiumtriacetat Hydrat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Terbium(III) acetate hydrate

A17388.06 A17388.14
A17388.06EA 138 EUR
A17388.06 A17388.14
Terbiumtriacetat Hydrat 99%
Terbiumtriacetat Hydrat

Purity: 99.99% CAS No.: 100587-92-6 Molecular Formula: Tb(OOCCH3)3.xH2O Formula Weight: 336.08(anhy) MDL No.: MFCD00149233

Formel: C₆H₉O₆Tb·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149233
CAS-Nummer: 100587-92-6
EINECS: 247-065-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR