Ytterbiumtriacetat Tetrahydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ytterbium(III) acetate tetrahydrate
M-Clarity™ Qualitätsstufe = MQ100
326011-10G
326011-50G
SIAL326011-10GEA
77.7
EUR
SIAL326011-10G
SIAL326011-50G
Ytterbiumtriacetat Tetrahydrat, Sigma-Aldrich®
Ytterbiumtriacetat Tetrahydrat
Formel:
C₆H₉O₆Yb·4H₂O Molecular Weight: 422,23 g/mol |
CAS-Nummer:
15280-58-7 EINECS: 244-137-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...