Ytterbiumtriacetat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ytterbium(III) acetate hydrate

M-Clarity™ Qualitätsstufe = MQ100

544973-25G 544973-5G
SIAL544973-25GEA 265 EUR
SIAL544973-25G SIAL544973-5G
Ytterbiumtriacetat Hydrat, Sigma-Aldrich®
Ytterbiumtriacetat Hydrat
Formel: C₆H₉O₆Yb·H₂O
Molecular Weight: 368,19 g/mol
MDL: MFCD00150127
CAS-Nummer: 15280-58-7
EINECS: 244-137-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR