Ytterbiumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific

Synonyme: Ytterbium(III) acetate hydrate

14576.09 14576.18
14576.09EA 66.5 EUR
14576.09 14576.18
Ytterbiumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Ytterbiumtriacetat Hydrat

Purity: 99.9% CAS No.: 15280-58-7 Molecular Formula: Yb(OOCCH3)3.xH2O Formula Weight: 350.16(anhy) Physical Form: Crystalline Aggregates Odour: Odourless MDL No.: MFCD00150127

Formel: C₆H₉O₆Yb·H₂O
Molecular Weight: 368,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150127
CAS-Nummer: 15280-58-7
EINECS: 244-137-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR