2,6-Dibrom-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

OT-0338-100G OT-0338-25G OT-0338-5G
COBBOT-0338-100GEA 127 EUR
COBBOT-0338-100G COBBOT-0338-25G COBBOT-0338-5G
2,6-Dibrom-4-(trifluormethoxy)anilin
2,6-Dibrom-4-(trifluormethoxy)anilin
Formel: C₇H₄Br₂F₃NO
MDL: MFCD00153113
CAS-Nummer: 88149-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR