(±)-Methyl-2-methylbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl2-methylbutyrate

M-Clarity™ Qualitätsstufe = MQ300

W271918-100G-K W271918-1KG-K W271918-20KG-K W271918-4KG-K
SIALW271918-100G-KEA 81 EUR
SIALW271918-100G-K SIALW271918-1KG-K SIALW271918-20KG-K SIALW271918-4KG-K
(±)-Methyl-2-methylbutyrat, Sigma-Aldrich®
(±)-Methyl-2-methylbutyrat
Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
MDL: MFCD00009335
CAS-Nummer: 868-57-5
EINECS: 212-778-0
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR