6-Brombenzothiazol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Bromobenzo[d]thiazole

H52823.MD H52823.03
H52823.MDEA 58.8 EUR
H52823.MD H52823.03
6-Brombenzothiazol ≥97%
6-Bromobenzothiazole

Purity: 97% CAS No.: 53218-26-1 Molecular Formula: C7H4BrNS Formula Weight: 214.08 Melting Point: 52-54° MDL No.: MFCD04115372

Formel: C₇H₄BrNS
Molecular Weight: 214.09 g/mol
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115372
CAS-Nummer: 53218-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR