4-Brom-2-(trifluormethyl)benzaldehyd

Lieferant: COMBI-BLOCKS

CA-4956-5G CA-4956-1G
COBBCA-4956-5GEA 0 EUR
COBBCA-4956-5G COBBCA-4956-1G
4-Brom-2-(trifluormethyl)benzaldehyd
4-Bromo-2-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
MDL: MFCD09037472
CAS-Nummer: 861928-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR