2-Amino-5-brombenzothiazol

Lieferant: COMBI-BLOCKS

OT-0937-5G OT-0937-25G OT-0937-1G
COBBOT-0937-5GEA 151 EUR
COBBOT-0937-5G COBBOT-0937-25G COBBOT-0937-1G
2-Amino-5-brombenzothiazol
2-Amino-5-bromobenzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229.1 g/mol
MDL: MFCD00577806
CAS-Nummer: 20358-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR