2-Amino-4,6-dichlorbenzothiazol

Lieferant: COMBI-BLOCKS

HC-6974-1G HC-6974-5G HC-6974-25G
COBBHC-6974-1GEA 65.8 EUR
COBBHC-6974-1G COBBHC-6974-5G COBBHC-6974-25G
2-Amino-4,6-dichlorbenzothiazol
2-Amino-4,6-dichlorobenzothiazole
Formel: C₇H₄Cl₂N₂S
MDL: MFCD01123002
CAS-Nummer: 16582-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR