2-Amino-4-chlorbenzothiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

133434-25G 133434-5G
SIAL133434-25GEA 45 EUR
SIAL133434-25G SIAL133434-5G
2-Amino-4-chlorbenzothiazol, Sigma-Aldrich®
2-Amino-4-chlorobenzothiazole
Formel: C₇H₅ClN₂S
Molecular Weight: 184,65 g/mol
MDL: MFCD00005791
CAS-Nummer: 19952-47-7
EINECS: 243-439-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR