Dinatriummalonathydrat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Malonic acid disodium salt hydrate

413500250 413501000 413505000
ACRO413500250EA 30.5 EUR
ACRO413500250 ACRO413501000 ACRO413505000
Dinatriummalonathydrat 99%
Dinatriummalonat Monohydrat
Formel: C₃H₂Na₂O₄·xH₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149114
CAS-Nummer: 132499-82-2
EINECS: 205-514-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder and/or chunks
Infrared spectrum Conforms
Water 7 to 13 % (K.F.)
Titration with HClO4 ≥98.5 % (On dry substance)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR