5-Fluorsalicylaldehyd

Lieferant: FLUOROCHEM

Synonyme: 5-Fluoro-2-hydroxybenzaldehyde

013016-1G 013016-25G 013016-5G
FLUO013016-1GEA 0 EUR
FLUO013016-1G FLUO013016-25G FLUO013016-5G
5-Fluorsalicylaldehyd
5-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,11 g/mol
MDL: MFCD01090997
CAS-Nummer: 347-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR