5-Fluorsalicylaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluoro-2-hydroxybenzaldehyde

43844.03 43844.06
43844.03EA 107 EUR
43844.03 43844.06
5-Fluorsalicylaldehyd ≥98%
5-Fluorsalicylaldehyd

Purity: 98+% CAS No.: 347-54-6 Molecular Formula: C7H5FO2 Formula Weight: 140.12 Physical Form: Powder/flakes Sensitivity: Air Sensitive MDL No.: MFCD01090997

Formel: C₇H₅FO₂
Molecular Weight: 140,11 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01090997
CAS-Nummer: 347-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR