2-Amino-5-fluorbenzothiazol 98%
Lieferant: Thermo Scientific
Synonyme:
5-Fluorobenzo[d]thiazol-2-amine
H51022.MD
H51022.03
H51022.MDEA
66.5
EUR
H51022.MD
H51022.03
2-Amino-5-fluorbenzothiazol 98%
2-Amino-5-fluorobenzothiazole
Purity: 98%
CAS No.: 20358-07-0
Molecular Formula: C7H5FN2S
Formula Weight: 168.19
MDL No.: MFCD05664560
Formel:
C₇H₅FN₂S Molecular Weight: 168,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664560 CAS-Nummer: 20358-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...