3,5-Bis(trifluormethyl)thiophenol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,5-Bis(trifluoromethyl)benzenethiol
A14669.03
A14669.06
A14669.03EA
73.4
EUR
A14669.03
A14669.06
3,5-Bis(trifluormethyl)thiophenol ≥98%
3,5-Bis(trifluormethyl)thiophenol
Purity: 98%
CAS No.: 130783-02-7
Molecular Formula: C8H4F6S
Formula Weight: 246.18
Boiling Point: 63-64°/15mm
UN No.: 2810
MDL No.: MFCD00042273
Formel:
C₈H₄F₆S Molecular Weight: 246,18 g/mol Siedepunkt: 63…64 °C (15 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042273 CAS-Nummer: 130783-02-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...