Tryptamin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(1H-indol-3-yl)ethan-1-amine
186120100
186120500
186122500
ACRO186120100EA
29.6
EUR
ACRO186120100
ACRO186120500
ACRO186122500
Tryptamin 98%
Tryptamin
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160,22 g/mol Siedepunkt: 137 °C (,1 mmHg) Schmelzpunkt: 113…117 °C Flammpunkt: 185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005661 CAS-Nummer: 61-54-1 EINECS: 200-510-5 |
Spezifikation Testergebnisse
Appearance | White to yellow-brown Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 113 to 117 °C |
Water | ≤2 % (K.F.) |
GC | ≥97.5 % |
Appearance of solution | (1% soln. in MeOH) clear, yellowish |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...