Tryptamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(1H-indol-3-yl)ethan-1-amine

M-Clarity™ Qualitätsstufe = MQ100

193747-10G 193747-50G
SIAL193747-10GEA 32.4 EUR
SIAL193747-10G SIAL193747-50G
Tryptamin, Sigma-Aldrich®
Tryptamin
Formel: C₁₀H₁₂N₂
Molecular Weight: 160.22 g/mol
MDL: MFCD00005661
CAS-Nummer: 61-54-1
EINECS: 200-510-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR