Tryptamin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(1H-indol-3-yl)ethan-1-amine
A11116.09
A11116.18
AB117522.0010
A11116.09EA
29.7
EUR
A11116.09
A11116.18
ABCRAB117522.0010
Tryptamin ≥98%
Tryptamin
Purity: 98+%
CAS No.: 61-54-1
Molecular Formula: C10H12N2
Formula Weight: 160.22
Storage Temperature: 2°C to 8°C
Flash Point: 185°(365°F)
Melting Point: 111-116°
Boiling Point: 137°/0.15mm
Beilstein Reference: 125513
Merck Reference: 14,9796
MDL No.: MFCD00005661
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160.22 g/mol Siedepunkt: 137 °C (0.15 mmHg) Schmelzpunkt: 111…116 °C Flammpunkt: 185 °C (365 °F) |
MDL:
MFCD00005661 CAS-Nummer: 61-54-1 EINECS: 200-510-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...