7-Benzyloxyindol

Lieferant: COMBI-BLOCKS

Synonyme: 7-(Benzyloxy)-1H-indole

IN-0058-5G IN-0058-25G IN-0058-1G
COBBIN-0058-5GEA 102 EUR
COBBIN-0058-5G COBBIN-0058-25G COBBIN-0058-1G
7-Benzyloxyindol
7-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
MDL: MFCD00037974
CAS-Nummer: 20289-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR