1-Butyl-3-methylimidazoliumoctylsulfat ≥99%
Lieferant: Thermo Scientific
Achtung
Synonyme: [BMIM] [OctSO4]
H27720.06 H27720.18
H27720.06EA 36.5 EUR
H27720.06 H27720.18
1-Butyl-3-methylimidazoliumoctylsulfat ≥99%
1-Butyl-3-methylimidazolium octylsulphate
Purity: 99% CAS No.: 445473-58-5 Molecular Formula: C16H32N2O4S Formula Weight: 348.50 MDL No.: MFCD03788914
Formel: C₁₆H₃₂N₂O₄S
Molecular Weight: 348,51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788914
CAS-Nummer: 445473-58-5