Hydroxyhydrochinontrimethylether ≥98%

Lieferant: Thermo Scientific

Synonyme: 1,2,4-Trimethoxybenzol

A10483.09 A10483.18
A10483.09EA 41.9 EUR
A10483.09 A10483.18
Hydroxyhydrochinontrimethylether ≥98%
Hydroxyhydrochinontrimethylether

Purity: 98% CAS No.: 135-77-3 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Refractive Index: 1.5330 Boiling Point: 247° Beilstein Reference: 2047579 Merck Reference: 14,1073 MDL No.: MFCD00008360

Formel: C₉H₁₂O₃
Molecular Weight: 168,19 g/mol
Siedepunkt: 247 °C
Schmelzpunkt: 19…21 °C
Dichte: 1,128
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008360
CAS-Nummer: 135-77-3
EINECS: 205-219-7
Merck Index: 14,1073

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR