Isoamylbutyrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Isopentyl butyrate, Isoamyl Butyrate
M-Clarity™ Qualitätsstufe = MQ300
W206008-1KG-K
W206008-20KG-K
W206008-9KG-K
SIALW206008-1KG-KEA
80.6
EUR
SIALW206008-1KG-K
SIALW206008-20KG-K
SIALW206008-9KG-K
Isoamylbutyrat, Sigma-Aldrich®
Isoamylbutyrat
Formel:
C₉H₁₈O₂ Molecular Weight: 158,24 g/mol |
MDL:
MFCD00044888 CAS-Nummer: 106-27-4 EINECS: 203-380-8 UN: 2620 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...