5-Brom-2-ethoxybenzaldehyd

Lieferant: FLUOROCHEM

011096-25G 011096-100G
FLUO011096-25GEA 350 EUR
FLUO011096-25G FLUO011096-100G
5-Brom-2-ethoxybenzaldehyd
5-Brom-2-ethoxybenzaldehyd
Formel: C₉H₉BrO₂
MDL: MFCD00016600
CAS-Nummer: 79636-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR