5-Brom-2-ethoxybenzaldehyd

Lieferant: MENAI

DH593.100 DH593.500 DH593.50
MENADH593.100EA 0 EUR
MENADH593.100 MENADH593.500 MENADH593.50
5-Brom-2-ethoxybenzaldehyd
5-Brom-2-ethoxybenzaldehyd
Formel: C₉H₉BrO₂
MDL: MFCD00016600
CAS-Nummer: 79636-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR