4-Brom-o-anisaldehyd
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-2-methoxybenzaldehyde
CA-4794-5G
CA-4794-1G
COBBCA-4794-5GEA
102
EUR
COBBCA-4794-5G
COBBCA-4794-1G
4-Brom-o-anisaldehyd
4-Brom-o-anisaldehyd
Formel:
C₈H₇BrO₂ Molecular Weight: 215.05 g/mol |
MDL:
MFCD08276823 CAS-Nummer: 43192-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...