2-Brom-5-chloranilin

Lieferant: COMBI-BLOCKS

AN-1056-5G AN-1056-25G
COBBAN-1056-5GEA 48.1 EUR
COBBAN-1056-5G COBBAN-1056-25G
2-Brom-5-chloranilin
2-Brom-5-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
MDL: MFCD00672942
CAS-Nummer: 823-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR