1-Brom-3,5-difluor-2-iodbenzol

Lieferant: FLUOROCHEM

004195-5G 004195-25G
FLUO004195-5GEA 42.9 EUR
FLUO004195-5G FLUO004195-25G
1-Brom-3,5-difluor-2-iodbenzol
1-Bromo-3,5-difluoro-2-iodobenzene
Formel: C₆H₂BrF₂I
Molecular Weight: 318.89 g/mol
MDL: MFCD00042183
CAS-Nummer: 175278-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR