3-Brom-2-fluoranilin

Lieferant: COMBI-BLOCKS

AN-1517-5G AN-1517-1G
COBBAN-1517-5GEA 166 EUR
COBBAN-1517-5G COBBAN-1517-1G
3-Brom-2-fluoranilin
3-Brom-2-fluoranilin
Formel: C₆H₅BrFN
MDL: MFCD09864700
CAS-Nummer: 58534-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR