4-Brom-2,3-difluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-2,3-difluoro-4-methoxybenzene

OT-0478-5G OT-0478-1G
COBBOT-0478-5GEA 117 EUR
COBBOT-0478-5G COBBOT-0478-1G
4-Brom-2,3-difluoranisol
4-Brom-2,3-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD07368017
CAS-Nummer: 406482-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR