3,5-Dibromanilin

Lieferant: COMBI-BLOCKS

AN-1206-5G AN-1206-25G AN-1206-1G
COBBAN-1206-5GEA 102 EUR
COBBAN-1206-5G COBBAN-1206-25G COBBAN-1206-1G
3,5-Dibromanilin
3,5-Dibromanilin
Formel: C₆H₅Br₂N
MDL: MFCD00047841
CAS-Nummer: 626-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR