3-Chlor-o-anisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chloro-2-methoxyaniline

M-Clarity™ Qualitätsstufe = MQ100

149942-1G 149942-5G
SIAL149942-1GEA 30.2 EUR
SIAL149942-1G SIAL149942-5G
3-Chlor-o-anisidin, Sigma-Aldrich®
3-Chlor-o-anisidin
Formel: C₇H₈ClNO
Molecular Weight: 157.6 g/mol
MDL: MFCD00060607
CAS-Nummer: 51114-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR