1-Boc-5-cyan-2-indolboronsäure

Lieferant: CHEMPUR

Synonyme: (1-(tert-Butoxycarbonyl)-5-cyano-1H-indol-2-yl)boronic acid, 5-Cyano-1H-indole-2-boronic acid, N-BOC protected

32-0014.1 32-0014.5
CHMP32-0014.1EA 0 EUR
CHMP32-0014.1 CHMP32-0014.5
1-Boc-5-cyan-2-indolboronsäure
1-Boc-5-cyan-2-indolboronsäure
Formel: C₁₄H₁₅BN₂O₄
MDL: MFCD04114578
CAS-Nummer: 475102-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR