1-Boc-6-cyan-2-indolboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Cyano-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-cyano-1H-indol-2-yl)boronic acid

H52520.MD H52520.03
H52520.MDEA 51.8 EUR
H52520.MD H52520.03
1-Boc-6-cyan-2-indolboronsäure 96%
1-Boc-6-cyan-2-indolboronsäure

Purity: 96% CAS No.: 913835-67-3 Molecular Formula: C14H15BN2O4 Formula Weight: 286.09 Storage Temperature: -30°C to -10°C Melting Point: 219-221° MDL No.: MFCD08436056

Formel: C₁₄H₁₅BN₂O₄
Molecular Weight: 286.09 g/mol
Schmelzpunkt: 219…221 °C
MDL: MFCD08436056
CAS-Nummer: 913835-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR