1-Boc-6-cyan-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Cyano-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-cyano-1H-indol-2-yl)boronic acid
BB-3467-1G
BB-3467-5G
COBBBB-3467-1GEA
243
EUR
COBBBB-3467-1G
COBBBB-3467-5G
1-Boc-6-cyan-2-indolboronsäure
1-Boc-6-cyan-2-indolboronsäure
Formel:
C₁₄H₁₅BN₂O₄ Molecular Weight: 286.09 g/mol |
MDL:
MFCD08436056 CAS-Nummer: 913835-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...