1-Boc-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
BB-2009-5G
BB-2009-25G
BB-2009-1G
COBBBB-2009-5GEA
137
EUR
COBBBB-2009-5G
COBBBB-2009-25G
COBBBB-2009-1G
1-Boc-2-indolboronsäure
1-Boc-2-indolboronsäure
Formel:
C₁₃H₁₆BNO₄ Molecular Weight: 261,08 g/mol |
MDL:
MFCD02093045 CAS-Nummer: 213318-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...