1-Boc-5-methoxy-2-indolboronsäure

Lieferant: CHEMPUR

Synonyme: N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid

32-0021.1 32-0021.5
CHMP32-0021.1EA 0 EUR
CHMP32-0021.1 CHMP32-0021.5
1-Boc-5-methoxy-2-indolboronsäure
1-Boc-5-methoxy-2-indolboronsäure
Formel: C₁₄H₁₈BNO₅
Molecular Weight: 291,11 g/mol
MDL: MFCD04039006
CAS-Nummer: 290331-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR