1-Boc-5-methoxy-2-indolboronsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid
H52654.03
H52654.06
H52654.03EA
48
EUR
H52654.03
H52654.06
1-Boc-5-methoxy-2-indolboronsäure 95%
1-Boc-5-methoxy-2-indolboronsäure
Purity: 95%
CAS No.: 290331-71-4
Molecular Formula: C14H18BNO5
Formula Weight: 291.11
Storage Temperature: -30°C to -10°C
Melting Point: 105-108°
MDL No.: MFCD04039006
Formel:
C₁₄H₁₈BNO₅ Molecular Weight: 291,11 g/mol Schmelzpunkt: 105…108 °C |
MDL:
MFCD04039006 CAS-Nummer: 290331-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...