1-Boc-6-methoxy-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Methoxy-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methoxy-1H-indol-2-yl)boronic acid
BB-2244-5G
BB-2244-1G
COBBBB-2244-5GEA
0
EUR
COBBBB-2244-5G
COBBBB-2244-1G
1-Boc-6-methoxy-2-indolboronsäure
1-Boc-6-methoxy-2-indolboronsäure
Formel:
C₁₄H₁₈BNO₅ Molecular Weight: 291,11 g/mol |
MDL:
MFCD06659830 CAS-Nummer: 850568-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...