1-Boc-7-methoxy-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 7-Methoxy-1H-indole-2-boronic acid, N-BOC protected

BB-3501-1G BB-3501-5G
COBBBB-3501-1GEA 375 EUR
COBBBB-3501-1G COBBBB-3501-5G
1-Boc-7-methoxy-2-indolboronsäure
1-Boc-7-methoxy-2-indolboronsäure
Formel: C₁₄H₁₈BNO₅
Molecular Weight: 291.11 g/mol
MDL: MFCD08689542
CAS-Nummer: 913835-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR