1-Boc-4-brom-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid, 4-Bromoindole-2-boronic acid, N-BOC protected
BB-3879-5G
BB-3879-1G
COBBBB-3879-5GEA
1660
EUR
COBBBB-3879-5G
COBBBB-3879-1G
1-Boc-4-brom-2-indolboronsäure
1-Boc-4-brom-2-indolboronsäure
Formel:
C₁₃H₁₅BBrNO₄ Molecular Weight: 339,98 g/mol |
MDL:
MFCD06801699 CAS-Nummer: 957034-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...