1-Boc-5-brom-2-indolboronsäure

Lieferant: CHEMPUR

Synonyme: (5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid, N-(Boc-5-bromo-2-indolyl)boronic acid, 5-Bromo-N-(Boc)indole-2-boronic acid

32-0018.1 32-0018.5
CHMP32-0018.1EA 0 EUR
CHMP32-0018.1 CHMP32-0018.5
1-Boc-5-brom-2-indolboronsäure
1-Boc-5-brom-2-indolboronsäure
Formel: C₁₃H₁₅BBrNO₄
Molecular Weight: 339.98 g/mol
MDL: MFCD03701683
CAS-Nummer: 475102-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR