1-Boc-6-brom-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid

BB-5313-5G BB-5313-1G
COBBBB-5313-5GEA 671 EUR
COBBBB-5313-5G COBBBB-5313-1G
1-Boc-6-brom-2-indolboronsäure
1-Boc-6-brom-2-indolboronsäure
Formel: C₁₃H₁₅BBrNO₄
Molecular Weight: 339.98 g/mol
MDL: MFCD15143453
CAS-Nummer: 1217500-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR