1-Boc-4-chlor-2-indolboronsäure

Lieferant: CHEMPUR

Synonyme: 4-Chloro-N-(Boc)-indole-2-boronic acid, (1-(tert-Butoxycarbonyl)-4-chloro-1H-indol-2-yl)boronic acid

32-0022.500 32-0022.1
CHMP32-0022.500EA 0 EUR
CHMP32-0022.500 CHMP32-0022.1
1-Boc-4-chlor-2-indolboronsäure
1-Boc-4-chlor-2-indolboronsäure
Formel: C₁₃H₁₅BClNO₄
Molecular Weight: 295.53 g/mol
MDL: MFCD04114582
CAS-Nummer: 475102-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR