1-Boc-5-chlor-2-indolboronsäure

Lieferant: CHEMPUR

Synonyme: (1-(tert-Butoxycarbonyl)-5-chloro-1H-indol-2-yl)boronic acid

32-0012.500 32-0012.1
CHMP32-0012.500EA 0 EUR
CHMP32-0012.500 CHMP32-0012.1
1-Boc-5-chlor-2-indolboronsäure
1-Boc-5-chlor-2-indolboronsäure
Formel: C₁₃H₁₅BClNO₄
Molecular Weight: 295,53 g/mol
MDL: MFCD04039005
CAS-Nummer: 475102-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR