1-Boc-6-chlor-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloroindole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-chloro-1H-indol-2-yl)boronic acid

BB-3548-1G BB-3548-5G
COBBBB-3548-1GEA 276 EUR
COBBBB-3548-1G COBBBB-3548-5G
1-Boc-6-chlor-2-indolboronsäure
1-Boc-6-chlor-2-indolboronsäure
Formel: C₁₃H₁₅BClNO₄
Molecular Weight: 295,53 g/mol
MDL: MFCD08689539
CAS-Nummer: 352359-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR