1-Boc-5-fluor-2-indolboronsäure
Lieferant: CHEMPUR
Synonyme:
1-Boc-5-fluoroindole-2-boronic Acid, N-(Boc)-5-fluoroindole-2-boronic acid
32-0020.1
32-0020.5
CHMP32-0020.1EA
0
EUR
CHMP32-0020.1
CHMP32-0020.5
1-Boc-5-fluor-2-indolboronsäure
1-Boc-5-fluor-2-indolboronsäure
Formel:
C₁₃H₁₅BFNO₄ Molecular Weight: 279.07 g/mol |
MDL:
MFCD04114581 CAS-Nummer: 352359-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...