1-Boc-4-methyl-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: N-(tert-Butoxycarbonyl)-4-methylindole-2-boronic acid

BB-9022-1G BB-9022-5G
COBBBB-9022-1GEA 276 EUR
COBBBB-9022-1G COBBBB-9022-5G
1-Boc-4-methyl-2-indolboronsäure
1-Boc-4-methyl-2-indolboronsäure
Formel: C₁₄H₁₈BNO₄
Molecular Weight: 275.11 g/mol
MDL: MFCD09265525
CAS-Nummer: 352359-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR