1-Boc-5-methyl-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(tert-Butoxycarbonyl)-5-methyl-1H-indol-2-yl)boronic acid, 5-Methyl-1H-indole-2-boronic acid, N-BOC protected

BB-2083-5G BB-2083-1G
COBBBB-2083-5GEA 1170 EUR
COBBBB-2083-5G COBBBB-2083-1G
1-Boc-5-methyl-2-indolboronsäure
1-Boc-5-methyl-2-indolboronsäure
Formel: C₁₄H₁₈BNO₄
Molecular Weight: 275.11 g/mol
MDL: MFCD04114580
CAS-Nummer: 475102-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR